This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RESP18 SIRNA Assays ›
Gene Symbol:
Resp18
Gene Name:
regulated endocrine-specific protein 18
Gene Aliases:
AI851012
Chromosome Location:
Chr.1: 75272197 - 75278455 on Build GRCm38
UniGene ID:
Mm.4032
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 64082

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Resp18
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009049.1 NP_033075.1 2 232
GenBank mRNA AK012477.1 - 2 185
AK019440.1 - 2 182
BC027535.1 - 2 195 3376899
FJ621497.1 - 2 168
L34214.1 - 2 232

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-5djdz:80/100.66.79.206:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline