siRNA Details
| Gene Symbol: Sepw1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009156.2 | NP_033182.1 | 4 | 228 | ||
| GenBank mRNA | AF015284.1 | - | 4 | 183 | ||
| AF241527.2 | - | 4 | 181 | |||
| AK002870.1 | - | 4 | 204 | |||
| AK028124.1 | - | 4 | 198 | |||
| BC052719.1 | - | 4 | 190 | 6814242 | ||
| BU604713.1 | - | 4 | 261 | |||
| U67890.1 | - | 4 | 180 | |||

Sepw1
Genomic Map