This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SH3BP1 SIRNA Assays ›
Gene Symbol:
Sh3bp1
Gene Name:
SH3-domain binding protein 1
Gene Aliases:
3BP-1
Chromosome Location:
Chr.15: 78899780 - 78912052 on Build GRCm38
UniGene ID:
Mm.4462
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 64454

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sh3bp1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009164.2 NP_033190.2 7 710
GenBank mRNA AK145156.1 - 7 650
AK161215.1 - 6 634
AK166337.1 - 7 703
AK172060.1 - 7 707
BC004598.1 - 5 431 3584958
X87671.1 - 4 566

Back To Top

More Information


Gene Ontology Categories:

Process(es)

SH3 domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-7wnc4:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline