This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SRM SIRNA Assays ›
Gene Symbol:
Srm
Gene Name:
spermidine synthase
Gene Aliases:
AA407669, SpdST, SpdSy
Chromosome Location:
Chr.4: 148591513 - 148594619 on Build GRCm38
UniGene ID:
Mm.10
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 64805

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Srm
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009272.4 NP_033298.1 7 1075
GenBank mRNA AK028359.1 - 7 1086
AK051101.1 - 7 1071
AK146395.1 - 7 1032
AK219795.1 - 1 86
BC005566.1 - 7 1079 3966710
L19311.1 - 7 1043

Back To Top

More Information


Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

polyamine metabolic process
polyamine biosynthetic process
spermidine biosynthetic process
catalytic activity
spermidine synthase activity
transferase activity
protein homodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-49kgl:80/100.66.79.151:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0