This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other UROS SIRNA Assays ›
Gene Symbol:
Uros
Gene Name:
uroporphyrinogen III synthase
Gene Aliases:
AI415298, UROIIIS, Uros3
Chromosome Location:
Chr.7: 133686142 - 133710519 on Build GRCm38
UniGene ID:
Mm.3160
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 65277

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Uros
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AA272198.1 - 3 288
AK135314.1 - 4 344
AK145902.1 - 4 394
AK148116.1 - 4 531
AK164297.1 - 4 309
BC068161.1 - 4 337 30358504
U04439.1 - 3 258
U16216.1 - 4 399
U18867.2 - 4 403

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001012068, XM_008759891, XM_008759892, XM_008759890, BC083730.

Panther Classification:

Molecular Function -

hydrolase metabolite interconversion enzyme

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline