This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BIRC5 SIRNA Assays ›
Gene Symbol:
Birc5
Gene Name:
baculoviral IAP repeat-containing 5
Gene Aliases:
AAC-11, Api4, TIAP, survivin40
Chromosome Location:
Chr.11: 117849237 - 117855743 on Build GRCm38
UniGene ID:
Mm.8552
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 65706

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Birc5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012273.1 NP_001012273.1 2 308
NM_009689.2 NP_033819.1 2 308
GenBank mRNA AB013819.1 - 2 302
AF077349.1 - 2 209
AK018022.1 - 1 530
AK142768.1 - 2 224
BC004702.1 - 2 226 3499203
BY023910.1 - 2 318
CN724527.1 - 2 308
EU366955.1 - 2 194

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s) Process(es)

G2/M transition of mitotic cell cycle
microtubule cytoskeleton organization
cytokinesis
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
apoptotic process
cell cycle
chromosome segregation
mitotic nuclear division
regulation of mitotic cell cycle
embryo development
regulation of signal transduction
negative regulation of peptidase activity
protein ubiquitination
protein complex localization
positive regulation of exit from mitosis
spindle checkpoint
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of neuron apoptotic process
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
cell division
establishment of chromosome localization
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of type B pancreatic cell proliferation
mitotic spindle assembly
inhibition of cysteine-type endopeptidase activity involved in apoptotic process
ubiquitin-protein transferase activity
cysteine-type endopeptidase inhibitor activity
protein binding
microtubule binding
zinc ion binding
Ran GTPase binding
tubulin binding
enzyme binding
peptidase inhibitor activity
identical protein binding
protein homodimerization activity
cysteine-type endopeptidase inhibitor activity involved in apoptotic process
metal ion binding
protein heterodimerization activity
cofactor binding
chaperone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline