siRNA Details
| Gene Symbol: Bcl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009744.3 | NP_033874.1 | 10 | 2281 | ||
| GenBank mRNA | AK036975.1 | - | 10 | 2305 | ||
| AK039228.1 | - | 10 | 2303 | |||
| AK151229.1 | - | 10 | 2306 | |||
| BC052315.1 | - | 10 | 2282 | 6309948 | ||
| U41465.1 | - | 9 | 2293 | |||

Bcl6
Genomic Map