siRNA Details
| Gene Symbol: Cst7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009977.3 | NP_034107.2 | 2 | 270 | ||
| GenBank mRNA | AB015224.1 | - | 2 | 251 | ||
| AF031825.1 | - | 2 | 199 | |||
| AK004420.1 | - | 2 | 271 | |||
| AK170583.1 | - | 2 | 217 | |||
| BC117047.1 | - | 2 | 255 | 40126366 | ||
| BC117049.1 | - | 2 | 255 | 40126368 | ||

Cst7
Genomic Map