This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CYP2C37 SIRNA Assays ›
Gene Symbol:
Cyp2c37
Gene Name:
cytochrome P450, family 2. subfamily c, polypeptide 37
Gene Aliases:
-
Chromosome Location:
Chr.19: 39992424 - 40012243 on Build GRCm38
UniGene ID:
Mm.38963
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 66412

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyp2c37
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF047542.1 - 827
BC057912.1 - 828 5052873

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline