This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ADAM28 SIRNA Assays ›
Gene Symbol:
Adam28
Gene Name:
a disintegrin and metallopeptidase domain 28
Gene Aliases:
C130072N01Rik, D430033C21Rik, Dtgn1, MDC-L, MDC-Lm, MDC-Ls, MDCL, TECADAM, eMDCII
Chromosome Location:
Chr.14: 68604994 - 68655887 on Build GRCm38
UniGene ID:
Mm.117450
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 66556

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Adam28
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048175.2 NP_001041640.1 2, 3 316
NM_010082.2 NP_034212.1 2, 3 316
NM_183366.3 NP_899222.1 2, 3 316
GenBank mRNA AF153350.1 - 2, 3 223
AF163290.1 - 2, 3 316
AF163291.1 - 2, 3 316
AF163292.1 - 2, 3 316
AF163293.1 - 2, 3 316
AK036800.1 - 2, 3 256
AK052479.1 - 2, 3 207
BC058782.1 - 2, 3 191 6310763

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_181693, XM_006252231, XM_008770820, AY283187.

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
metalloendopeptidase activity
peptidase activity
metallopeptidase activity
zinc ion binding
hydrolase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline