This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other OPN3 SIRNA Assays ›
Gene Symbol:
Opn3
Gene Name:
opsin 3
Gene Aliases:
ERO, Ecpn
Chromosome Location:
Chr.1: 175662421 - 175692902 on Build GRCm38
UniGene ID:
Mm.32744
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 66662

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Opn3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010098.3 NP_034228.1 2 631
GenBank mRNA AF140241.1 - 2 589
AK043135.1 - 2 632
AK132822.1 - 2 568
BC104486.1 - 3 481
BC104487.1 - 3 504
BC105646.1 - 3 428
BC105919.1 - 3 428
BC139805.1 - 3 429

Back To Top

More Information


Set Membership:

G-protein coupled receptor

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
phototransduction
protein-chromophore linkage
response to stimulus
signal transducer activity
G-protein coupled receptor activity
photoreceptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-ftbch:80/100.66.79.106:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0