siRNA Details
| Gene Symbol: Fbln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010180.2 | NP_034310.2 | 17 | 2539 | ||
| GenBank mRNA | AK085989.1 | - | 17 | 2484 | ||
| AK137511.1 | - | 17 | 2486 | |||
| AK159646.1 | - | 17 | 2486 | |||
| AK159816.1 | - | 6 | 976 | |||
| X70854.1 | - | 17 | 2527 | |||

Fbln1
Genomic Map