siRNA Details
| Gene Symbol: Sprr2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011470.2 | NP_035600.1 | 2 | 328 | ||
| GenBank mRNA | AJ005562.1 | - | 2 | 310 | ||
| AK008778.1 | - | 2 | 329 | |||
| AY158988.1 | - | 2 | 328 | |||
| BC111436.1 | - | 2 | 309 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr2d| Gene Symbol: Sprr2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011470.2 | NP_035600.1 | 2 | 328 | ||
| GenBank mRNA | AJ005562.1 | - | 2 | 310 | ||
| AK008778.1 | - | 2 | 329 | |||
| AY158988.1 | - | 2 | 328 | |||
| BC111436.1 | - | 2 | 309 | |||