siRNA Details
| Gene Symbol: Sprr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011472.2 | NP_035602.1 | 2 | 570 | ||
| GenBank mRNA | AJ005564.1 | - | 1 | 531 | ||
| AY158990.1 | - | 2 | 570 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sprr2f| Gene Symbol: Sprr2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011472.2 | NP_035602.1 | 2 | 570 | ||
| GenBank mRNA | AJ005564.1 | - | 1 | 531 | ||
| AY158990.1 | - | 2 | 570 | |||