This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TRY5 SIRNA Assays ›
Gene Symbol:
Try5 Try4
Gene Name:
trypsin 5
trypsin 4
Gene Aliases:
1810049H19Rik, Tc, Tesp4
0910001B19Rik, Td
Chromosome Location:
Chr.6: 41311232 - 41314710 on Build GRCm38
Chromosome Location:
Chr.6: 41302272 - 41305533 on Build GRCm38
UniGene ID:
Mm.333022
Mm.383263
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 69664

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Try5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003405.4 NP_001003405.1 3 328
GenBank mRNA AB009661.1 - 3 328
AK007843.1 - 3 327
AK131898.1 - 3 327
AK160484.1 - 3 327
BC139220.1 - 3 413 8862242
BC139223.1 - 3 413 8862245
Gene Symbol: Try4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011646.5 NP_035776.1 3 326
GenBank mRNA AK003064.1 - 3 330
AK008660.1 - 3 327
AK008667.1 - 3 327
BC050120.1 - 3 307
BC061135.1 - 3 326 30306436

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
serine-type endopeptidase activity
calcium ion binding
serine-type peptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rp79v:80/100.66.77.211:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline