This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SKP1A SIRNA Assays ›
Gene Symbol:
Skp1a
Gene Name:
S-phase kinase-associated protein 1A
Gene Aliases:
15kDa, 2610043E24Rik, 2610206H23Rik, EMC19, OCP-II, OCP2, SKP1, Tceb1l, p19A, p19Skp1
Chromosome Location:
Chr.11: 52230522 - 52246858 on Build GRCm38
UniGene ID:
Mm.42944
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 69775

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Skp1a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011543.4 NP_035673.3 4 351
GenBank mRNA AF083214.1 - 4 275
AK002983.1 - 4 322
AK010628.1 - 4 337
AK012498.1 - 4 310
AK014245.1 - 4 310
AK027909.1 - 4 309
AK078330.1 - 4 337
AK088339.1 - 4 310
AK152609.1 - 4 309
AK160198.1 - 4 310
AK166673.1 - 4 310
BC002115.1 - 4 294 3490159
BU841654.1 - 5 443
Z47088.1 - 4 300

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

ubiquitin-dependent protein catabolic process
protein ubiquitination
SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
histone H2A monoubiquitination
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline