This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other XLR3A SIRNA Assays ›
Gene Symbol:
Xlr3a
Gene Name:
X-linked lymphocyte-regulated 3A
Gene Aliases:
Xlr3
Chromosome Location:
Chr.X: 73086293 - 73097095 on Build GRCm38
UniGene ID:
Mm.475098
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 69911

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Xlr3a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110784.1 NP_001104254.1 5, 6 457
GenBank mRNA BC003812.1 - 5, 6 403 3601144
BC093495.1 - 380 3593889
BC107255.1 - 366 40052405
BC107283.1 - 367 40052875
BC127145.1 - 367 40052876
BC127146.1 - 368 40052877
BQ885410.1 - 5, 6 397
DV662513.1 - 5, 6 405
U02599.1 - 4, 5 350

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008759621, XM_006229630, XM_008759622, XM_006229628.

Gene Ontology Categories:

Function(s) Process(es)

spermatid development
biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4kmk2:80/100.66.78.54:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline