This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GADD45G SIRNA Assays ›
Gene Symbol:
Gadd45g
Gene Name:
growth arrest and DNA-damage-inducible 45 gamma
Gene Aliases:
AI327420, C86281, CR6, DDIT2, OIG37
Chromosome Location:
Chr.13: 51846675 - 51848475 on Build GRCm38
UniGene ID:
Mm.281298
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 70349

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gadd45g
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011817.2 NP_035947.2 3 338
GenBank mRNA AB021884.1 - 3 335
AF055638.1 - 3 326
AK002237.1 - 3 336
AK007410.1 - 3 341
BC001989.1 - 3 319 3493618

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

activation of MAPKKK activity
activation of MAPKK activity
negative regulation of protein kinase activity
apoptotic process
response to stress
multicellular organism development
cell differentiation
interferon-gamma biosynthetic process
positive regulation of apoptotic process
T-helper 1 cell differentiation
positive regulation of JNK cascade
regulation of cell cycle
positive regulation of p38MAPK cascade
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-vzlfm:80/100.66.78.148:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0