This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NLRP5 SIRNA Assays ›
Gene Symbol:
Nlrp5 Nlrp5-ps
Gene Name:
NLR family, pyrin domain containing 5
NLR family, pyrin domain containing 5, pseudogene
Gene Aliases:
Mater, Nalp5, Op1, PAN11
Gm18756
Chromosome Location:
Chr.7: 23385897 - 23441921 on Build GRCm38
Chromosome Location:
Chr.7: 14530654 - 14623066 on Build GRCm38
UniGene ID:
Mm.333653
Mm.372763
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 70385

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nlrp5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF074018.1 - 9 2299
AK087774.1 - 9 2321
AY196361.1 - 9 2299
AY196362.1 - 9 2299
AY329484.1 - 8 2251
AY329485.1 - 9 2299
AY329486.1 - 9 2299
AY329487.1 - 8 2251
AY329488.1 - 9 2320
AY329489.1 - 9 2299
AY329490.1 - 9 2299
AY329491.1 - 9 2320
BC053384.1 - 10 2366 30049556
Gene Symbol: Nlrp5-ps
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK016782.1 - 7 748
AK140342.1 - 7 995
BC150849.1 - 5 448

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-sp9vq:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline