This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ST6GALNAC5 SIRNA Assays ›
Gene Symbol:
St6galnac5
Gene Name:
ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases:
AI851940, Siat7e
Chromosome Location:
Chr.3: 152820709 - 152982207 on Build GRCm38
UniGene ID:
Mm.40915
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 70899

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: St6galnac5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012028.4 NP_036158.3 5 1886
GenBank mRNA AB028840.1 - 1869
AK034387.1 - 1890
AK038434.1 - 5 1890
BC055737.1 - 5 1886 6415191

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
glycosphingolipid biosynthetic process
oligosaccharide biosynthetic process
sialylation
alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase activity
sialyltransferase activity
lytic endotransglycosylase activity
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-xwmhz:80/100.66.76.181:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0