This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GRCC10 SIRNA Assays ›
Gene Symbol:
Grcc10
Gene Name:
gene rich cluster, C10 gene
Gene Aliases:
2310033H05Rik, C10
Chromosome Location:
Chr.6: 124739183 - 124741079 on Build GRCm38
UniGene ID:
Mm.439877
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 71001

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Grcc10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013535.1 NP_038563.1 2 308
GenBank mRNA AK009591.1 - 2 235
AK013899.1 - 2 293
BC005683.1 - 2 287 3154555

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001198725, XM_006244638, XM_003750659, XM_006237325, FM062242.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline