siRNA Details
| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF022363.1 | - | 6 | 3459 | ||
| S79832.1 | - | 6 | 2511 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp1| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF022363.1 | - | 6 | 3459 | ||
| S79832.1 | - | 6 | 2511 | |||