This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BLCAP SIRNA Assays ›
Gene Symbol:
Blcap
Gene Name:
bladder cancer associated protein homolog (human)
Gene Aliases:
AI462828, Bc10
Chromosome Location:
Chr.2: 157556362 - 157566952 on Build GRCm38
UniGene ID:
Mm.480461
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 72821

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Blcap
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016916.3 NP_058612.3 2 1011
GenBank mRNA AB037891.1 - 2 966
AK011673.1 - 2 988
AK018127.1 - 2 1041
AK170338.1 - 2 988
BC026935.1 - 2 975 3597231
BC083339.1 - 2 975 6832329

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133582, BC087661.

Gene Ontology Categories:

Function(s)

apoptotic process
cell cycle
apoptotic nuclear changes

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0