This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RAB11A SIRNA Assays ›
Gene Symbol:
Rab11a
Gene Name:
RAB11A, member RAS oncogene family
Gene Aliases:
-
Chromosome Location:
Chr.9: 64715300 - 64737756 on Build GRCm38
UniGene ID:
Mm.1387
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 73151

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rab11a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017382.5 NP_059078.2 5 812
GenBank mRNA AK014268.1 - 5 813
AK031143.1 - 5 715
AK132072.1 - 5 796
AK132159.1 - 5 798
AK147122.1 - 5 813
BC010722.1 - 5 801 2811973
D50500.1 - 3 320

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_004663, NM_001206836, X56740, BC013348, AF000231, and others.

Gene Ontology Categories:

Function(s) Process(es)

cytokinesis
transport
intracellular protein transport
exocytosis
cell cycle
mitotic metaphase plate congression
small GTPase mediated signal transduction
metabolic process
positive regulation of epithelial cell migration
regulation of multivesicular body size
positive regulation of G2/M transition of mitotic cell cycle
protein transport
vesicle-mediated transport
astral microtubule organization
neuron projection development
melanosome transport
Rab protein signal transduction
multivesicular body assembly
positive regulation of axon extension
regulation of long-term neuronal synaptic plasticity
regulation of protein transport
establishment of vesicle localization
regulation of vesicle-mediated transport
establishment of protein localization to organelle
protein localization to plasma membrane
establishment of protein localization to membrane
mitotic spindle assembly
exosomal secretion
nucleotide binding
GTPase activity
protein binding
GTP binding
microtubule binding
myosin V binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-qcwr4:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline