This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CYHR1 SIRNA Assays ›
Gene Symbol:
Cyhr1
Gene Name:
cysteine and histidine rich 1
Gene Aliases:
1110031M01Rik, AU042374, Chrp
Chromosome Location:
Chr.15: 76643395 - 76660208 on Build GRCm38
UniGene ID:
Mm.247752
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 73611

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Cyhr1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001276321.1 NP_001263250.1 2, 3 789
NM_019396.3 NP_062269.1 3, 4 722
GenBank mRNA AJ251516.1 - 3, 4 575
AK080867.1 - 2, 3 810
AK129156.1 - 1, 2 1896
BC039646.1 - 1, 2 115
BC062184.1 - 2, 3 778 6515695
BC082772.1 - 2, 3 789 6508580
BE653118.1 - 1, 2 148
BX512375.1 - 2, 3 328
BY287678.1 - 2, 3 353
CF742880.1 - 1, 2 237

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_002729828, BC168705.

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline