This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NUDT3 SIRNA Assays ›
Gene Symbol:
Nudt3
Gene Name:
nudix (nucleotide diphosphate linked moiety X)-type motif 3
Gene Aliases:
1110011B09Rik, AA960325, Dipp, Dipp1
Chromosome Location:
Chr.17: 27579382 - 27623452 on Build GRCm38
UniGene ID:
Mm.144699
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 74397

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Nudt3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291046.1 NP_001277975.1 3 506
NM_019837.2 NP_062811.1 3 498
GenBank mRNA AF264064.1 - 3 497
BC016534.1 - 4 404 2615904
BC046805.1 - 3 499 6829170
BU919402.1 - 3 330

Back To Top

More Information


Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

diphosphoinositol polyphosphate catabolic process
magnesium ion binding
diphosphoinositol-polyphosphate diphosphatase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-596jt:80/100.66.79.70:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline