This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDCD10 SIRNA Assays ›
Gene Symbol:
Pdcd10
Gene Name:
programmed cell death 10
Gene Aliases:
2410003B13Rik, Ccm3, Tfa15, Tfar15
Chromosome Location:
Chr.3: 75516490 - 75556852 on Build GRCm38
UniGene ID:
Mm.316473
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 74502

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdcd10
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019745.4 NP_062719.2 5, 6 658
GenBank mRNA AF159368.1 - 5, 6 561
AK005594.1 - 5, 6 587
AK075926.1 - 5, 6 606
AK077308.1 - 5, 6 635
AK137198.1 - 5, 6 628
AK149182.1 - 5, 6 564
AK160234.1 - 5, 6 608
AK168857.1 - 5, 6 592
AK169689.1 - 5, 6 592
BC019650.1 - 5, 6 598 4022513
BC086778.1 - 5, 6 577 5689088

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_145860, NM_145859, NM_007217, AK291130, BC016353, and others.
This siRNA also targets the following transcripts in Rat: NM_001009542, XM_006232501, FQ213019, BC067245.

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
apoptotic process
positive regulation of cell proliferation
positive regulation of gene expression
negative regulation of gene expression
positive regulation of cell migration
positive regulation of stress-activated MAPK cascade
positive regulation of peptidyl-serine phosphorylation
intrinsic apoptotic signaling pathway in response to hydrogen peroxide
response to hydrogen peroxide
negative regulation of apoptotic process
positive regulation of MAP kinase activity
wound healing, spreading of cells
positive regulation of Notch signaling pathway
protein stabilization
establishment of Golgi localization
positive regulation of protein serine/threonine kinase activity
negative regulation of cell migration involved in sprouting angiogenesis
Golgi reassembly
negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
protein binding
protein kinase binding
protein homodimerization activity
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline