This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SELK SIRNA Assays ›
Gene Symbol:
Selk BC052688 Gm12060
Gene Name:
selenoprotein K
cDNA sequence BC052688
selenoprotein K pseudogene
Gene Aliases:
1110001C03Rik, AA673253, HSPC030, Hsp30
-
OTTMUSG00000005275
Chromosome Location:
Chr.14: 29968380 - 29975074 on Build GRCm38
Chromosome Location:
Chr.13: 61771580 - 61787282 on Build GRCm38
Chromosome Location:
Chr.11: 23558058 - 23558842 on Build GRCm38
UniGene ID:
Mm.104491
Mm.317358
Mm.247328
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
siRNA ID 74850

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Selk
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019979.2 NP_064363.2 4, 5 427
GenBank mRNA AF170920.1 - 4, 5 456
AK003206.1 - 4, 5 355
AK003280.1 - 21
AK006674.1 - 4, 5 327
AK007523.1 - 4, 5 339
AK008419.1 - 4, 5 363
BC038049.1 - 4, 5 368 2645414
BC096044.1 - 4, 5 309 1248280
BC096677.1 - 4, 5 357 874920
Gene Symbol: BC052688
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC066226.1 - 2720
Gene Symbol: Gm12060
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC027278.1 - 1 401

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
calcium ion transport
response to oxidative stress
macrophage derived foam cell differentiation
protein palmitoylation
positive regulation of cell migration
positive regulation of T cell proliferation
respiratory burst after phagocytosis
regulation of protein transport
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
positive regulation of T cell migration
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline