siRNA Details
| Gene Symbol: Supt20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019995.3 | NP_064379.1 | 7 | 522 | ||
| GenBank mRNA | AF139179.1 | - | 7 | 526 | ||
| AK050619.1 | - | 8 | 546 | |||
| AK083379.1 | - | 8 | 546 | |||
| AK188434.1 | - | 5 | 326 | |||
| AK210270.1 | - | 4 | 306 | |||
| BC052702.1 | - | 7 | 511 | 6400922 | ||

Supt20
Genomic Map