This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DNAJC4 SIRNA Assays ›
Gene Symbol:
Dnajc4
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 4
Gene Aliases:
2010301J22Rik, Hspf2, Mcg18
Chromosome Location:
Chr.19: 6987911 - 6992696 on Build GRCm38
UniGene ID:
Mm.10721
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 75251

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dnajc4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020566.1 NP_065591.1 3 303
GenBank mRNA AF036875.1 - 3 303
AK008507.1 - 3 549
AK170008.1 - 3 542
BC049281.1 - 3 534 4989485

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001013196, XM_006230859, XM_006230861, XM_006230862, XM_006230860, and others.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-v7nth:80/100.66.79.110:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0