This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SULT5A1 SIRNA Assays ›
Gene Symbol:
Sult5a1
Gene Name:
sulfotransferase family 5A, member 1
Gene Aliases:
Sult-x1, Sultx1
Chromosome Location:
Chr.8: 123142843 - 123158914 on Build GRCm38
UniGene ID:
Mm.6562
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 75334

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Sult5a1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020564.3 NP_065589.2 4 492
GenBank mRNA AF026074.1 - 4 480
AK142243.1 - 4 493
BC012677.1 - 4 488 4187578
CJ065192.1 - 4 493

Back To Top

More Information


Panther Classification:

Molecular Function -

transferase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
molecular_function
heparan sulfate 2-O-sulfotransferase activity
HNK-1 sulfotransferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline