This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RHOX4A SIRNA Assays ›
Gene Symbol:
Rhox4a Rhox4b Rhox4c Rhox4d Rhox4e Rhox4f Rhox4g
Gene Name:
reproductive homeobox 4A
reproductive homeobox 4B
reproductive homeobox 4C
reproductive homeobox 4D
reproductive homeobox 4E
reproductive homeobox 4F
reproductive homeobox 4G
Gene Aliases:
Rhox4, Rhox4.1
5430432L21Rik, Ehox, Rhox4.2, Rhox4a
Rhox4.3, Rhox4b
Rhox4.4, Rhox4c
Rhox4, Rhox4.5, Rhox4d
Rhox4.6, Rhox4e, Rhox4h
Rhox4.6, Rhox4.7, Rhox4f
Chromosome Location:
Chr.X: 37265318 - 37270136 on Build GRCm38
Chromosome Location:
Chr.X: 37432494 - 37437278 on Build GRCm38
Chromosome Location:
Chr.X: 37480337 - 37485124 on Build GRCm38
Chromosome Location:
Chr.X: 37514389 - 37519176 on Build GRCm38
Chromosome Location:
Chr.X: 37557412 - 37562282 on Build GRCm38
Chromosome Location:
Chr.X: 37602895 - 37607686 on Build GRCm38
Chromosome Location:
Chr.X: 37646501 - 37651327 on Build GRCm38
UniGene ID:
Mm.296330
Mm.327419
Mm.296444
Mm.381921
Mm.379498
Mm.491809
Mm.491809
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
-
-
-
-
-
siRNA ID 75502

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Rhox4a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039688.3 NP_001034777.1 2, 3 612
GenBank mRNA AJ972665.1 - 2, 3 612
AK017380.1 - 2, 3 527
BC057925.1 - 2, 3 527 5292192
BC116370.1 - 2, 3 560 40100097
BC116371.1 - 2, 3 559 40100098
BC145382.1 - 2, 3 513 9053940
BC171959.1 - 2, 3 513 9054245
DQ058641.1 - 2, 3 587
Gene Symbol: Rhox4b
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021300.2 NP_067275.1 2, 3 607
GenBank mRNA AF265350.1 - 2, 3 595
AJ972666.1 - 2, 3 607
BC172126.1 - 2, 3 554 9054796
Gene Symbol: Rhox4c
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039689.1 NP_001034778.1 2, 3 624
GenBank mRNA AJ972667.1 - 2, 3 624
Gene Symbol: Rhox4d
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039695.1 NP_001034784.1 480
GenBank mRNA AB221641.1 - 476
AJ972668.1 - 480
BC147696.1 - 2, 3 554 9056669
Gene Symbol: Rhox4e
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201236.3 NP_957688.2 2, 3 698
GenBank mRNA AJ972669.1 - 2, 3 698
BC094891.1 - 2, 3 660 30019143
BC172128.1 - 554 9054798
Gene Symbol: Rhox4f
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039696.1 NP_001034785.1 2, 3 650
GenBank mRNA AJ972670.1 - 2, 3 650
Gene Symbol: Rhox4g
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039698.1 NP_001034787.1 2, 3 699
GenBank mRNA AJ972671.1 - 2, 3 699

Back To Top

More Information


Additional Information:

This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.

Gene Ontology Categories:

Function(s)

biological_process

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline