siRNA Details
| Gene Symbol: Scel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022886.2 | NP_075024.2 | 3 | 236 | ||
| GenBank mRNA | AF245700.2 | - | 3 | 236 | ||
| AK020346.1 | - | 3 | 158 | |||
| BC111874.1 | - | 3 | 156 | 40059586 | ||
| BC112380.1 | - | 3 | 157 | 40059585 | ||
| BY775767.1 | - | 3 | 237 | |||

Scel
Genomic Map