This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TNMD SIRNA Assays ›
Gene Symbol:
Tnmd
Gene Name:
tenomodulin
Gene Aliases:
1110017I01Rik, Bricd4, ChM1L, TeM
Chromosome Location:
Chr.X: 133851008 - 133865577 on Build GRCm38
UniGene ID:
Mm.46221
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 76380

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Tnmd
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022322.2 NP_071717.1 4, 5 615
GenBank mRNA AB055422.1 - 4, 5 422
AF191768.1 - 4, 5 561
AF219993.1 - 4, 5 488
AF291655.1 - 4, 5 640
AK014761.1 - 4, 5 454
BC006919.1 - 4, 5 424 3601223
BC049944.1 - 4, 5 615 2811628

Back To Top

More Information


Gene Ontology Categories:

Function(s)

endothelial cell morphogenesis
negative regulation of endothelial cell proliferation
negative regulation of angiogenesis

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-5mz9h:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0