This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2410016O06RIK SIRNA Assays ›
Gene Symbol:
2410016O06Rik
Gene Name:
RIKEN cDNA 2410016O06 gene
Gene Aliases:
MAPJD, NO66
Chromosome Location:
Chr.12: 83950608 - 83952953 on Build GRCm38
UniGene ID:
Mm.251483
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 77289

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2410016O06Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023633.3 NP_076122.2 1 1718
GenBank mRNA AB041553.1 - 1 1657
AK010521.1 - 1 824
AK080870.1 - 1 1664
AK213032.1 - 1 305
BC052152.2 - 1 1632
BC059809.1 - 1 1669
BC138094.1 - 1 1718 8861113

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108040.

Gene Ontology Categories:

Function(s) Process(es)

chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
histone H3-K4 demethylation
ribosome biogenesis
negative regulation of osteoblast differentiation
negative regulation of transcription, DNA-templated
oxidation-reduction process
histone H3-K36 demethylation
sulfonate dioxygenase activity
RNA binding
iron ion binding
protein binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, 2-oxoglutarate as one donor, and incorporation of one atom each of oxygen into both donors
2,4-dichlorophenoxyacetate alpha-ketoglutarate dioxygenase activity
procollagen-proline dioxygenase activity
histone demethylase activity (H3-K4 specific)
hypophosphite dioxygenase activity
DNA-N1-methyladenine dioxygenase activity
metal ion binding
dioxygenase activity
histone demethylase activity (H3-K36 specific)
C-19 gibberellin 2-beta-dioxygenase activity
C-20 gibberellin 2-beta-dioxygenase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-47mq8:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline