siRNA Details
| Gene Symbol: Fam120b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024203.3 | NP_077165.2 | 2, 3 | 1708 | ||
| GenBank mRNA | AK016568.1 | - | 2, 3 | 1674 | ||
| AK132211.1 | - | 2, 3 | 1686 | |||
| BC003200.1 | - | 1, 2 | 611 | |||
| BC066088.1 | - | 2, 3 | 1753 | |||
| DQ873694.1 | - | 2, 3 | 1689 | |||

Fam120b
Genomic Map