siRNA Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025388.2 | NP_079664.1 | 2 | 240 | ||
| GenBank mRNA | AK003070.1 | - | 1 | 142 | ||
| AK003897.1 | - | 2 | 241 | |||
| AK011260.1 | - | 1 | 131 | |||
| BC021936.1 | - | 2 | 201 | 5124904 | ||

Ufc1
Genomic Map