siRNA Details
| Gene Symbol: Nicn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025449.3 | NP_079725.1 | 2 | 220 | ||
| GenBank mRNA | AJ299741.1 | - | 2 | 212 | ||
| AK002789.1 | - | 2 | 226 | |||
| AK013602.1 | - | 2 | 248 | |||
| AK075732.1 | - | 2 | 228 | |||
| AK146890.1 | - | 2 | 230 | |||
| AK169135.1 | - | 2 | 230 | |||
| BC024050.1 | - | 2 | 220 | 5133763 | ||

Nicn1
Genomic Map