siRNA Details
| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025520.3 | NP_079796.1 | 5 | 295 | ||
| GenBank mRNA | AK008480.1 | - | 4 | 553 | ||
| AK009617.1 | - | 5 | 289 | |||
| AK011218.1 | - | 5 | 281 | |||
| BC048459.1 | - | 5 | 270 | 6601124 | ||
| BC061085.1 | - | 279 | 30295556 | |||
| BC132626.1 | - | 5 | 295 | 40130903 | ||
| BC132628.1 | - | 5 | 295 | 40130905 | ||
| CB204385.1 | - | 5 | 288 | |||

Lsm5
Genomic Map