This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 2610528J11RIK SIRNA Assays ›
Gene Symbol:
2610528J11Rik
Gene Name:
RIKEN cDNA 2610528J11 gene
Gene Aliases:
TEMP
Chromosome Location:
Chr.4: 118526995 - 118530217 on Build GRCm38
UniGene ID:
Mm.34431
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 78466

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 2610528J11Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025572.2 NP_079848.1 3 411
GenBank mRNA AK009128.1 - 3 410
AK012175.1 - 3 444
AK147115.1 - 3 409
BC022764.1 - 3 434 4925508

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_001164829, NM_182517, AL696898, BC041633.

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-245sx:80/100.66.79.25:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0