This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1700029H14RIK SIRNA Assays ›
Gene Symbol:
1700029H14Rik
Gene Name:
RIKEN cDNA 1700029H14 gene
Gene Aliases:
-
Chromosome Location:
Chr.8: 13550722 - 13562554 on Build GRCm38
UniGene ID:
Mm.46136
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 78584

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 286.00
Your Price
Online offer: 277.65
277.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1700029H14Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080781.2 NP_001074250.1 2 326
NM_025601.3 NP_079877.1 2 326
GenBank mRNA AK006340.1 - 2 271
AK006493.1 - 2 269
AK016674.1 - 2 271
BC049575.1 - 2 292 6773883
CA464636.1 - 2 316

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline