This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CTRB1 SIRNA Assays ›
Gene Symbol:
Ctrb1
Gene Name:
chymotrypsinogen B1
Gene Aliases:
2200008D09Rik, AI504462, Ctrb, Prt-2
Chromosome Location:
Chr.8: 111686510 - 111691010 on Build GRCm38
UniGene ID:
Mm.34374
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 78663

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ctrb1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025583.2 NP_079859.2 4 323
GenBank mRNA AK003060.1 - 4 332
AK003079.1 - 4 324
AK007566.1 - 4 323
AK007765.1 - 4 323
AK007815.1 - 4 324
AK008644.2 - 4 324
AK008729.1 - 4 324
AK008888.1 - 4 324
AK008927.1 - 4 324
AK018554.1 - 4 325
BC061083.1 - 4 323 30316047

Back To Top

More Information


Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
digestion
protein catabolic process
response to cytokine
positive regulation of apoptotic process
serine-type endopeptidase activity
peptidase activity
serine-type peptidase activity
hydrolase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-q76w2:80/100.66.77.41:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0