siRNA Details
| Gene Symbol: Slc16a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025807.3 | NP_080083.2 | 2 | 293 | ||
| GenBank mRNA | AK004556.1 | - | 2 | 294 | ||
| AK004599.1 | - | 2 | 472 | |||
| AK004684.2 | - | 2 | 351 | |||
| BC034205.1 | - | 2 | 248 | 4987055 | ||
| BC055839.1 | - | 2 | 251 | 4237811 | ||

Slc16a9
Genomic Map