siRNA Details
| Gene Symbol: Slc16a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025807.3 | NP_080083.2 | 5 | 624 | ||
| GenBank mRNA | AK004556.1 | - | 5 | 625 | ||
| AK004599.1 | - | 5 | 803 | |||
| AK004684.2 | - | 5 | 682 | |||
| AK015196.1 | - | 3 | 401 | |||
| BC034205.1 | - | 5 | 579 | 4987055 | ||
| BC055839.1 | - | 5 | 582 | 4237811 | ||

Slc16a9
Genomic Map