siRNA Details
| Gene Symbol: Use1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145780.1 | NP_001139252.1 | 9 | 1042 | ||
| NM_025917.4 | NP_080193.1 | 8 | 829 | |||
| GenBank mRNA | AF353245.1 | - | 8 | 822 | ||
| AK003257.1 | - | 8 | 822 | |||
| AK003385.1 | - | 7 | 693 | |||
| AK008573.1 | - | 8 | 822 | |||
| AK008661.1 | - | 7 | 727 | |||
| AK012490.1 | - | 8 | 823 | |||
| AK017484.1 | - | 5 | 896 | |||
| AK160726.1 | - | 8 | 822 | |||
| AK165639.1 | - | 8 | 971 | |||
| BC025086.1 | - | 7 | 905 | 3995001 | ||
| BC038519.1 | - | 7 | 892 | 1428854 | ||
| BC075695.1 | - | 9 | 948 | 30623689 | ||

Use1
Genomic Map