This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other F5 SIRNA Assays ›
Gene Symbol:
F5
Gene Name:
coagulation factor V
Gene Aliases:
FVL, PCCF, RPRGL1, THPH2
Chromosome Location:
Chr.1: 169511954 - 169586632 on Build GRCh38
UniGene ID:
Hs.30054
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 8005

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: F5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000130.4 NP_000121.2 13 4668
XM_017000660.1 XP_016856149.1 12 4677
GenBank mRNA AJ297255.1 - 1 2548
AK226079.1 - 13 4586
M14335.1 - 13 4599
M16967.1 - 13 4613

Back To Top

More Information


Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

platelet degranulation
ER to Golgi vesicle-mediated transport
blood coagulation
blood circulation
platelet activation
COPII vesicle coating
copper ion binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline