siRNA Details
| Gene Symbol: Mri1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026423.4 | NP_080699.3 | 2 | 462 | ||
| GenBank mRNA | AK007709.1 | - | 2 | 423 | ||
| AK010538.1 | - | 2 | 434 | |||
| AK013882.1 | - | 2 | 431 | |||
| AK047287.1 | - | 2 | 465 | |||
| AK144807.1 | - | 2 | 403 | |||
| AK164635.1 | - | 2 | 462 | |||
| AK164894.1 | - | 2 | 431 | |||
| BC025049.1 | - | 2 | 429 | 4951045 | ||

Mri1
Genomic Map