This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DALRD3 SIRNA Assays ›
Gene Symbol:
Dalrd3
Gene Name:
DALR anticodon binding domain containing 3
Gene Aliases:
6330580J24Rik, C77829, mir-425
Chromosome Location:
Chr.9: 108567496 - 108572773 on Build GRCm38
UniGene ID:
Mm.458081
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 80510

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Dalrd3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026378.3 NP_080654.3 9 1285
GenBank mRNA AK002439.1 - 9 1281
AK003374.1 - 3 232
AK149963.1 - 9 1286
AK169405.1 - 9 1283
AK171818.1 - 9 1286
BC013483.1 - 9 1266 4242135
BC058774.1 - 7 684

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
arginyl-tRNA aminoacylation
biological_process
nucleotide binding
aminoacyl-tRNA ligase activity
arginine-tRNA ligase activity
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline