This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other 1810043G02RIK SIRNA Assays ›
Gene Symbol:
1810043G02Rik
Gene Name:
RIKEN cDNA 1810043G02 gene
Gene Aliases:
AV026620, D10Jhu13e
Chromosome Location:
Chr.10: 77978530 - 77985438 on Build GRCm38
UniGene ID:
Mm.297512
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 80643

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: 1810043G02Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026431.2 NP_080707.2 6, 7 668
GenBank mRNA AK003097.1 - 6, 7 676
AK004032.1 - 6, 7 670
AK075794.1 - 6, 7 655
AK165293.1 - 3, 4 3469
BC016605.1 - 5, 6 1393

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
smoothened signaling pathway
regulation of cell shape
cell projection organization
cilium morphogenesis
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0