siRNA Details
| Gene Symbol: Emc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026519.3 | NP_080795.1 | 3 | 402 | ||
| GenBank mRNA | AK003668.1 | - | 3 | 370 | ||
| AK012047.1 | - | 3 | 403 | |||
| AK078165.1 | - | 3 | 403 | |||
| AK167745.1 | - | 3 | 354 | |||
| BC051926.1 | - | 3 | 358 | 5710720 | ||

Emc4
Genomic Map